Learn More
Invitrogen™ Phospho-Ezrin/Radixin/Moesin (Thr567, Thr564, Thr558) Recombinant Rabbit Monoclonal Antibody (PSH05-73)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA556581
Artikelnummer. 30276201
  • 467.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Positive Control: HeLa starved for 3 hours then add 100 nM Calyculin A for 30 minutes cell lysate, NIH/3T3 starved for 24 hours then add 100 nM Calyculin A for 30 minutes cell lysate, C6 treated with 100 ng/mL Calyculin A for 1 hours cell lysate, mouse liver tissue, rat liver tissue. Tissue Specificity: Low tissue specificity. Subcellular Location: Cell membrane. Cell projection. Cytoplasm. Cytoskeleton. Membrane. Sequence Similarities: 100% Mouse/Rat. Predicted band size: 69 kDa.
Moesin, a member of the talin-4.1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Phospho-Ezrin/Radixin/Moesin (Thr567, Thr564, Thr558)
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
O35763, P15311, P26038, P26040, P26041, P26043, P31977, P35241
Ezr, MSN, RDX
Synthetic phosphopeptide corresponding to residues surrounding Thr567 of human ezrin protein.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
PSH05-73
Unconjugated
Ezr
AA516625; AW146364; C78546; CVIL; CVL; cytovillin; cytovillin 2; DFNB24; epididymis luminal protein 70; epididymis secretory protein Li 105; ESP10; EZR; Ezrin; HEL70; HEL-S-105; membrane-organizing extension spike protein; Moesin; MSN; p81; R75297; radixin; radixin isoform; RDX; VIL2; villin 2; villin 2 (ezrin); villin-2
Rabbit
Protein A
RUO
17698, 19684, 22350, 315655, 4478, 54319, 5962, 7430, 81521
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.