missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-mTOR (Ser2448) Monoclonal Antibody (MRRBY), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

174.00 € - 425.00 €

Spezifikation

Antigen Phospho-mTOR (Ser2448)
Klon MRRBY
Konzentration 5 μL/Test
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15507226
Unterlagen ansehen
Invitrogen™
12-9718-41
25 Tests
174.00 €
25 Tests
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15517226
GREENER_CHOICE
Unterlagen ansehen
Invitrogen™
12-9718-42
100 Tests
425.00 €
100 Tests
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

Description: This MRRBY monoclonal antibody recognizes human and mouse mammalian target of rapamycin (also known as mTOR, FRAP, RAFT) when phosphorylated on S2448. mTOR is a serine/threonine protein kinase that functions as an ATP and amino acid sensor as well as to balance nutrient availability with cell growth, proliferation, motility, survival, protein synthesis, and transcription. Activated mTOR increases production of enzymes necessary for glycolysis and controls the uptake of glucose and other nutrients. Increased glucose uptake and metabolism helps fulfill the energy needs for mTOR-driven cell growth and proliferation. When sufficient nutrients are available, mTOR transmits a positive signal to p70 S6 kinase and participates in the inactivation of the eIF4E inhibitor, 4E-BP1. mTOR is phosphorylated at S2448 via the PI3 kinase/Akt signaling pathway and is autophosphorylated at Ser2481. Due to its critical role in regulation of cell growth, survival, and metabolism, and because it is often abnormally regulated in tumors, mTOR is under investigation as a potential target for anti-cancer therapy. Applications Reported: This MRRBY antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MRRBY antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test.

FRAP1 (mTOR) is a serine/threonine kinase that plays a critical role in cellular growth and proliferation. Perturbations in the mTOR/PI3-kinase/AKT pathway are associated with numerous forms of cancer. FRAP1 is also the target of rapamycin and its analogues, which are currently used as immunosuppressants and cancer therapeutics. Mutations affecting the gene results in Smith-Kingsmore syndrome.
Spezifikation

Spezifikation

Phospho-mTOR (Ser2448)
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
2610315D21Rik; AI327068; angiopoietin-like factor CDT6; FK506 binding protein 12-rapamycin associated protein 1; FK506 binding protein 12-rapamycin associated protein 2; FK506-binding protein 12-rapamycin complex-associated protein 1; FKBP12-rapamycin complex-associated protein; FKBP12-rapamycin complex-associated protein 1; FKBP-rapamycin associated protein; FKBP-rapamycin associated protein (FRAP); FKBP-rapamycin-associated protein FRAP; flat; FRAP; FRAP1; FRAP2; Mammalian target of rapamycin; mechanistic target of rapamycin; mechanistic target of rapamycin (serine/threonine kinase); mechanistic target of rapamycin kinase; Mtor; m-TOR; mTORC1; Raft1; Rapamycin and FKBP12 target 1; rapamycin and FKBP12 target-1 protein; rapamycin associated protein FRAP2; rapamycin target protein 1; RAPT1; serine/threonine-protein kinase mTOR; SKS
MTOR
Primary
4°C, store in dark, DO NOT FREEZE!
MTOR
MRRBY
Flow Cytometry
PE
Mouse
Human, Mouse
P42345, Q9JLN9
2475, 56717
IgG2a κ
Affinity chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.