missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PKC delta/theta (Ser643, Ser676) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA517905
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the phosphorylated PKC isoforms alpha, beta, gamma or epsilon.
PRKCD (Protein Kinase C Delta) is a Protein Coding gene. Diseases associated with PRKCD includeAutoimmune Lymphoproliferative Syndrome, Type IiiandSystemic Lupus Erythematosus 16. Among its related pathways areRole of Calcineurin dependent NFAT signaling in lymphocytesandWnt Signaling Pathway and Pluripotency. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus containing groupsandprotein tyrosine kinase activity. An important paralog of this gene isPRKCQ (PKC theta).
Spezifikation
| Phospho-PKC delta/theta (Ser643, Ser676) | |
| Polyclonal | |
| Unconjugated | |
| PRKCQ | |
| A130035A12Rik; AI385711; ALPS3; AW494342; CVID9; D14Ertd420e; may1; npkc-delta; nPKC-theta; PKC theta; PKC[d]; PKC-0; Pkcd; PKCd1; PKCd2; PKCdelta; PKC-delta; PKCdelta1; pkc-delta1; PKCdelta2; Pkcq; PKCtheta; PKC-theta; Prkcd; prkcd.L; prkcd.S; prkcd-a; prkcd-b; PRKCQ; PRKCT; protein kinase C delta; protein kinase C delta S homeolog; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C theta; protein kinase C theta type; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta L homeolog; protein kinase C, delta V; protein kinase C, theta; protein kinase C[d]; protein kinase delta1; SDK1; SDK-1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD; XELAEV_18025733mg | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 170538, 18753, 18761, 404226, 5580, 5588, 85420 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 15 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P09215, P28867, Q02111, Q04759, Q05655, Q9WTQ0 | |
| PRKCD, prkcd.S, PRKCQ | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer643 of rat PKCdelta. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Frog | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur