missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Progesterone Receptor (Ser190) Monoclonal Antibody (1154)

Artikelnummer. 10786263
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
1 Stück
Artikelnummer. Menge unitSize
10786263 100 μL 1 Stück
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10786263

Marke: Invitrogen™ 378200

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur für Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Mouse Monoclonal Antibody

37-8200 detects Phospho-Progesterone Receptor in Human samples. 37-8200 has been successfully used in western blot, immunofluorescence and immunohistochemistry in human samples. Phospho-Progesterone Receptor (Ser 190) antibody detects both progesterone receptor A and B forms when phosphorylated at Ser26 and Ser190, respectively.

The progesterone receptor (PR) is a member of the steroid family of nuclear receptors. The PR mediates the physiological effects of progesterone, which plays a central role in reproductive events associated with the establishment and maintenance of pregnancy. PR is found as a 94 kDa protein (Form A) or a 120 kDa protein (Form B) due to the use of alternative translation initiation sites. PR-B is the transcriptionally active form and is responsible for activating genes for the maintenance of the endometrium, maintenance of pregnancy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. In its inactive state, PgR forms a multiprotein complex which includes heat shock proteins and immunophins. Upon binding of progesterone hormone to its receptor, there is a conformational change that allows dimerization and binding of the receptor to progesterone response elements (PRE) sequences, resulting in activated transcription. A Null mutation in the PGR gene leads to pleiotrophic reproductive abnormalities.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Phospho-Progesterone Receptor (Ser190)
Anwendungen Immunohistochemistry, Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon 1154
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gen PGR
Gen-Zugriffsnummer P06401
Gen-Alias 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Gensymbole PGR
Wirtsspezies Mouse
Immunogen Synthetic phospho peptide derived from a region of the human Progesterone Receptor (hPR) protein surrounding the phosphorylated Ser190 residue.
Reinigungsverfahren Protein G
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 5241
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG1
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.