missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Tau (Thr217) Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 10536653
Ansicht ändern
Click to view available options
Menge:
100 μL
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
10536653 100 μL 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur f\u00FCr Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Artikelnummer. 10536653 Lieferant Invitrogen™ Lieferanten-Nr. 44744
Proteomik vorantreiben
Verwenden Sie den Aktionscode 28003
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Nur f\u00FCr Forschungszwecke. Die Anwendung muss den Produktanweisungen entsprechen.

Rabbit Polyclonal Antibody

Phospho-Tau (Thr217) Polyclonal Antibody for IHC (P), Western Blot

Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Phospho-Tau (Thr217)
Anwendungen Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Zusammensetzung Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
Gen MAPT
Gen-Zugriffsnummer P10636, P10637, P19332
Gen-Alias AI413597; AW045860; DDPAC; FLJ31424; FTDP17; FTDP-17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; Mapt; MAPTL; MGC138549; microtubule associated protein tau; microtubule-associated protein tau; microtubule-associated protein tau, isoform 4; microtubules; MSTD; Mtapt; MTBT1; MTBT2; Neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHFtau; PHF-tau; PPND; PPP1R103; protein phosphatase 1, regulatory subunit 103; pTau; RNPTAU; Tau; Tau microtubule-associated protein; tau protein; Tau-4; Tau5; Unknown (protein for MGC:134287)
Gensymbole MAPT
Wirtsspezies Rabbit
Immunogen The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human tau that contains threonine 217.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 17762, 29477, 4137
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.