missing translation for 'onlineSavingsMsg'
Learn More

ARC/NOL3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Artikelnummer. 18605179 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18605179 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605179 Supplier Novus Biologicals Supplier No. NBP241753JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARC/NOL3 Polyclonal antibody specifically detects ARC/NOL3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARC/NOL3
Anwendungen Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Janelia Fluor 549
Zusammensetzung 50mM Sodium Borate
Gen-Alias Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa
Wirtsspezies Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the carboxy terminus of human ARC. The immunogen is located within the last 50 amino acids of ARC. Amino Acid Squence: PDPEPEPDFEERDESEDS
Reinigungsverfahren Peptide affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis
Primär oder sekundär Primary
Gen-ID (Entrez) 8996
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.