Description
This gene encodes a phosphatidylinositol (PI) 4-kinase which catalyzes the first committed step in the biosynthesis of phosphatidylinositol 4,5-bisphosphate. The mammalian PI 4-kinases have been classified into two types, II and III, based on their molecular mass, and modulation by detergent and adenosine. The protein encoded by this gene is a type III enzyme that is not inhibited by adenosine. Two transcript variants encoding different isoforms have been described for this gene.
Specifications
Specifications
| Antigen | PI4KA |
| Anwendungen | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.17 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | PI4KA |
| Gen-Zugriffsnummer | E9Q3L2, O08662, P42356 |
| Gen-Alias | PI4 kinase alpha, PI4K ALPHA, pi4K230, PI4KA, PIK4, PIK4CA, PtdIns 4 kinase alpha |
| Gensymbole | PI4KA |
| Show More |