missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piezo1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15939-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Piezo1 is a pore-forming subunit of a mechanosensitive non-specific cation channel, and generates currents characterized by a linear current-voltage relationship that are sensitive to ruthenium red and gadolinium. Structurally, Piezo1 contains 36 transmembrane domains and functions as a homotetramer. Piezo1 plays a key role in epithelial cell adhesion by maintaining integrin activation through R-Ras recruitment to the ER, most probably in its activated state, and subsequent stimulation of calpain signaling. In the kidney, Piezo1 may contribute to the detection of intraluminal pressure changes and to urine flow sensing. Piezo1 acts as shear-stress sensor that promotes endothelial cell organization and alignment in the direction of blood flow through calpain activation and plays a key role in blood vessel formation and vascular structure in both development and adult physiology. The protein encoded by the Piezo1 gene is a mechanically-activated ion channel that links mechanical forces to biological signals. Defects in this gene have been associated with dehydrated hereditary stomatocytosis. Diseases associated with PIEZO1 include Dehydrated Hereditary Stomatocytosis with/without Pseudohyperkalemia And/Or Perinatal Edema and Lymphedema.Spezifikation
| Piezo1 | |
| Polyclonal | |
| Unconjugated | |
| PIEZO1 | |
| FAM38A, KIAA0233, Mib, PIEZO1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234839, 9780 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| E2JF22, Q92508 | |
| PIEZO1 | |
| Piezo1 Fusion Protein Ag7791 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur