missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pimivalimab Recombinant Monoclonal Antibody
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Pimivalimab, also known as JTX-4014, is a promising anti-PD-1 inhibitor primarily employed in oncology treatments. This biosimilar, located in the immunoglobulin superfamily (IgSF), is strategically designed to target the PD-1 receptor found on T-cells. The gene PDCD1, also known as PD-1 or CD279, encodes Programmed Cell Death Protein 1, a crucial immune checkpoint receptor expressed on T cells and other immune cells. Located on chromosome 2q37.3, PDCD1 consists of five exons. PD-1 is a type I transmembrane protein within the CD28/CTLA-4 superfamily and functions primarily to inhibit immune responses, thus maintaining peripheral tolerance and preventing autoimmune reactions. Upon binding to its ligands, PD-L1 and PD-L2, expressed on tumor cells and other tissues, PD-1 mediates the suppression of T-cell activity, inducing apoptosis in antigen-specific T cells while sparing regulatory T cells, contributing significantly to immune evasion by tumors.
Spezifikation
Spezifikation
| Antigen | Pimivalimab |
| Anwendungen | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Klassifikation | Recombinant Monoclonal |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Wirtsspezies | Human |
| Reinigungsverfahren | Protein A |
| Menge | 100 μg |
| Regulatorischer Status | RUO |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?