missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirimicarb-d6 (Dimethyl-d6-carbamoyl), CDN
Spezifikation
Spezifikation
| Chemischer Name oder Material | Pirimicarb D6 (dimethylcarbamate D6) |
| CAS | 1015854-66-6 |
| Summenformel | C11 2H6 H12 N4 O2 |
| Menge | 0.005 g |
| InChI-Formel | InChI=1 S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3/i5D3,6D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C(=O)Oc1nc(nc(C)c1C)N(C)C)C([2 H])([2 H])[2 H] |
| IUPAC-Name | [2-(Dimethylamino)-5,6-dimethylpyrimidin-4-yl] N,N-bis(trideuteriomethyl)carbamat |
| Molekulargewicht (g/mol) | 244.32 |
| Formelmasse | 244,1806 g/mol |
| Reinheit (%) | 99 Atom % D, Minimum 98% chemische Reinheit |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?