Learn More
Pirimicarb, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Pirimicarb |
| CAS | 23103-98-2 |
| Summenformel | C11 H18 N4 O2 |
| Menge | 250 mg |
| Synonym | Carbamic acid','dimethyl-','2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester (8CI,9CI), 4-Pyrimidinol','2-(dimethylamino)-5,6-dimethyl-','dimethylcarbamate (ester) (8CI), 2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate, 2-Dimethylamino-5,6-dimethylpyrimidin-4-yl N,N-dimethylcarbamate, 5,6-Dimethyl-2-dimethylamino-4-dimethylcarbamoyloxypyrimidine, Aficida, Aphox, Aphox DG, Fernos, PP 062, Patton Flow, Pirimicarb, Pirimix, Pirimor, Pirimor G, Pirimor Granulate, Pyrimor, Carbamic acid','N,N-dimethyl-','2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester |
| InChI-Formel | InChI=1S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3 |
| SMILES | CN(C)C(=O)Oc1nc(nc(C)c1C)N(C)C |
| IUPAC-Name | [2-(dimethylamino)-5,6-dimethylpyrimidin-4-yl] N,N-dimethylcarbamate |
| Molekulargewicht (g/mol) | 238.29 |
| Formelmasse | 238.143 |