Learn More
Invitrogen™ PKC zeta Recombinant Rabbit Monoclonal Antibody (24GB4195)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556938
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PKC zeta Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
Spezifikation
| PKC zeta | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKCZ | |
| 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; PKC2; Pkcz; PKC-ZETA; Prkcz; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, zeta; r14-3-3; R74924; zetaPKC | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18762, 25522, 5590 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4195 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P09217, Q02956, Q05513 | |
| PRKCZ | |
| A synthesized peptide derived from human PKC zeta (aa 350-592). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |