missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.50 €
Spezifikation
| Antigen | PKR |
|---|---|
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5-5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5-5 μg/mL |
| Anwendungen | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18772944
|
Novus Biologicals
NBP1-77266-0.025mg |
0.025 mg |
189.00 € 178.50 € / 0.03 Milligramm Sparen 10.50 € 5% Rabatt |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | |||||
Beschreibung
PKR Polyclonal specifically detects PKR in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Spezifikation
| PKR | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5-5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5-5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated | |
| EIF2AK2 | |
| IgG | |
| Affinity Purified | |
| Human PKR has 2 isoforms, including isoform 1 (551aa, 62.1kD) and isoform S (510aa, 57.4kD. This antibody detects both human isoforms. Mouse PKR has 1 isoform (515aa, 58.3kD). Rat PKR has only one isoform identified so far (513aa, 58.3kD). |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| AAP57628 | |
| 5610 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PKR. The immunogen is located within the last 50 amino acids of PKR. Amino Acid Squence: TVWKKSPEKNERHT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 68 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts