missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA596336
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: AVWQFRKMIK CVIPGSDPFL EYNNYGCYCG LGGSGTPVDE LDKCCQTHDN CYDQAKKLDS CKFLLDNPYT HTYSYSCSGS AITCSSKNKE CEAFICNCDR NAAICFSKAP YNKAHKNLDT KKYCQS; Positive Samples: A-549, Mouse pancreas, Mouse small intestine.
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.
Spezifikation
| PLA2 | |
| Polyclonal | |
| Unconjugated | |
| PLA2G1B | |
| Group IB phospholipase A2; group IB secretory phospholipase A(2); MGC119834; MGC119835; phosphatidylcholine 2-acylhydrolase 1B; phospholipase; phospholipase A2; phospholipase A2 group IB; phospholipase A2, group 1B; phospholipase A2, group IB (pancreas); phospholipase A2, group IB, pancreas; phospholipase A2, major isoenzyme; PLA2; PLA2A; Pla2g1b; PLA2-Ib; PPLA2; sPLA(2)-IB; sPLA2IB; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18778, 5319 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P04054, Q9Z0Y2 | |
| PLA2G1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-148 of human PLA2G1B (NP_0009191). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur