missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Placental Lactogen/CSH1 Polyclonal specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for ELISA/Immunohistochemistry/Immunohistochemistry (Paraffin).
Spezifikation
Spezifikation
| Antigen | Placental Lactogen/CSH1 |
| Anwendungen | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | PE |
| Verdünnung | ELISA, Immunohistochemistry-Paraffin |
| Zusammensetzung | PBS |
| Gen-Alias | Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin |
| Wirtsspezies | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Placental Lactogen/CSH1 (Accession#: NP_001308.1; Met1-Phe217) |
| Reinigungsverfahren | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?