Learn More
Invitrogen™ Plasminogen Kringle 5 Monoclonal Antibody (120304)
GREENER_CHOICE
Rat Monoclonal Antibody
Marke:  Invitrogen™ MA561813
Artikelnummer. 30536153
  • 606.00 € / 500 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
Plasminogen Kringle 5 is a domain within the plasminogen gene, which encodes the precursor to plasmin, an important enzyme involved in the breakdown of blood clots (fibrinolysis). The plasminogen protein contains multiple kringle domains, each contributing to its function. Kringle 5, in particular, has garnered interest due to its potent anti-angiogenic properties, meaning it can inhibit the formation of new blood vessels. This function is crucial in regulating processes such as wound healing, tissue regeneration, and the suppression of tumor growth, as tumors often rely on angiogenesis to obtain nutrients and grow. Kringle 5 exerts its effects by binding to specific receptors on endothelial cells, thereby inhibiting their proliferation and migration. This makes it a promising target for therapeutic strategies aimed at treating diseases characterized by excessive angiogenesis, such as cancer and certain eye diseases like diabetic retinopathy and age-related macular degeneration. Understanding the mechanisms of action and regulatory pathways involving Plasminogen Kringle 5 is essential for developing novel anti-angiogenic therapies and improving clinical outcomes in angiogenesis-related disorders.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Plasminogen Kringle 5
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
E. coli-derived recombinant mouse Plasminogen Kringle 5, Cys481-Ala563.
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
120304
Unconjugated
Rat
Protein A/G