missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plasminogen Kringle 5 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5149354
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Plasminogen Kringle 5 is a domain within the plasminogen gene, which encodes the precursor to plasmin, an important enzyme involved in the breakdown of blood clots (fibrinolysis). The plasminogen protein contains multiple kringle domains, each contributing to its function. Kringle 5, in particular, has garnered interest due to its potent anti-angiogenic properties, meaning it can inhibit the formation of new blood vessels. This function is crucial in regulating processes such as wound healing, tissue regeneration, and the suppression of tumor growth, as tumors often rely on angiogenesis to obtain nutrients and grow. Kringle 5 exerts its effects by binding to specific receptors on endothelial cells, thereby inhibiting their proliferation and migration. This makes it a promising target for therapeutic strategies aimed at treating diseases characterized by excessive angiogenesis, such as cancer and certain eye diseases like diabetic retinopathy and age-related macular degeneration. Understanding the mechanisms of action and regulatory pathways involving Plasminogen Kringle 5 is essential for developing novel anti-angiogenic therapies and improving clinical outcomes in angiogenesis-related disorders.
Spezifikation
| Plasminogen Kringle 5 | |
| Polyclonal | |
| Unconjugated | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| E. coli-derived recombinant mouse Plasminogen Kringle 5, Cys481-Ala563. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur