Learn More
Ibidi μ-Plate 24 Well Black ID 14 mm ibiTreat
Artikelnummer. 17331339
Encompass_Preferred
Change view
Click to view available options
Wells:
24
Packungsgröße:
15 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Wells unitSize
17331339 24 15 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17331339 Lieferant Ibidi Lieferanten-Nr. 82426
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
μ-Plate 24 Well Black ID 14 mm ibiTreat is a black 24 well plate with flat and clear bottom.
  • A black 24 well plate with flat and clear bottom for high throughput applications in cell-based assays
  • well ID: 14 mm
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Wells 24
Farbe Black
Durchmesser (metrisch) 14 mm