missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleiotrophin/PTN Antibody (CL15628) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-44293
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Pleiotrophin/PTN Monoclonal antibody specifically detects Pleiotrophin/PTN in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Spezifikation
| Pleiotrophin/PTN | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin | |
| This antibody was generated using a synthetic peptide of P21246, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5764 | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL15628 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur