missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLTP Polyclonal Antibody

Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5148932
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
PLTP is one of at least two lipid transfer proteins found in human plasma. The protein transfers phospholipids from triglyceride-rich lipoproteins to high density lipoprotein (HDL). In addition to regulating the size of HDL particles, this protein may be involved in cholesterol metabolism.
Spezifikation
| PLTP | |
| Polyclonal | |
| Unconjugated | |
| PLTP | |
| BPI fold containing family E; BPIFE; HDLCQ9; lipid transfer protein II; OD107; Phospholipid transfer protein; Pltp | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 18830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P55065 | |
| PLTP | |
| Mouse myeloma cell line NS0-derived recombinant mouse PLTP, Glu18-Ala493 (Asp32Glu). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Spot an opportunity for improvement?Share a Content Correction