missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (PDPN/4009R), Allophycocyanin, Novus Biologicals™
Alle ansehen Bio Techne ProdukteDescription
Podoplanin Monoclonal antibody specifically detects Podoplanin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Podoplanin |
| Anwendungen | ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | PDPN/4009R |
| Konjugat | APC |
| Zusammensetzung | PBS |
| Gen-Alias | 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha |
| Wirtsspezies | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues within aa1-100 of Podoplanin (Uniprot: Q86YL7) |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?