Beschreibung
The protein encoded by this gene is a zinc finger protein that may act as a transcription factor. The encoded protein may be part of a BHC histone deacetylase complex. Translocation of this gene with the fibroblast growth factor receptor-1 gene (FGFR1) results in a fusion gene, which may be a cause of stem cell leukemia lymphoma syndrome (SCLL). Several transcript variants encoding the same protein have been found for this gene.
Spezifikation
Spezifikation
| Antigen | POLA2 |
| Anwendungen | Immunofluorescence, Immunocytochemistry, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.39 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | POLA2 |
| Gen-Zugriffsnummer | Q14181 |
| Gen-Alias | DNA polymerase alpha subunit B, POLA2 |
| Gensymbole | POLA2 |
| Mehr anzeigen |