Learn More
Invitrogen™ POLB Recombinant Rabbit Monoclonal Antibody (6M8I5)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA535359
Beschreibung
Immunogen sequence: MGVCQLPSKN DEKEYPHRRI DIRLIPKDQY YCGVLYFTGS DIFNKNMRAH ALEKGFTINE YTIRPLGVTG VAGEPLPVDS EKDIFDYIQW KYREPKDRSE.
DNA polymerase beta comprises an amino-terminal 8- kDa domain and a carboxy-terminal 31- kDa domain. The N-terminal ssDNA binding domain has a deoxyribose phosphodiesterase activity while the C-terminal domain has a nucleotidyltransferase activity. Mammalian DNA polymerase beta, a DNA repair polymerase, is constitutively expressed in cultured cells, but treatment of cells with the DNA-alkylating agents such as N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) or methyl methanesulfonate up-regulates beta-pol level. DNA polymerase beta fills single nucleotide gaps in DNA produced by the base excision repair pathway of mammalian cells.
Spezifikation
| POLB | |
| Recombinant Monoclonal | |
| 0.22 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06746, P06766, Q8K409 | |
| POLB | |
| A synthetic peptide corresponding to a sequence within amino acids 236-335 of human DNA Polymerase beta (P06746). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6M8I5 | |
| Unconjugated | |
| POLB | |
| A430088C08Rik; DNA pol beta; DNA polymerase beta; DNA polymerase beta subunit; DNA-directed DNA polymerase beta; hypothetical protein LOC614688; LOW QUALITY PROTEIN: DNA polymerase beta; MGC125976; POL B; Pol beta; POLB; polb.S; polymerase (DNA directed), beta; polymerase (DNA directed), beta S homeolog; polymerase (DNA) beta; polymerase B; polymerase-beta; wu:fa97a05; XELAEV_18020165mg; zgc:109924 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18970, 29240, 5423 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.