Learn More
POLI Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Beschreibung
POLI Monoclonal antibody specifically detects POLI in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | POLI |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | OTI1H9 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gen | Rasgrp3 |
| Gen-Zugriffsnummer | Q8IV61 |
| Gen-Alias | DNA polymerase iota; eta2; Poli; polymerase (DNA directed) iota; polymerase (DNA directed), iota; polymerase (DNA) iota; polymerase (DNA-directed), iota; RAD 30B; RAD30; RAD30 homolog B; RAD30B; RAD3OB |
| Mehr anzeigen |