Learn More
POLI Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Artikelnummer. 17509842
Encompass_Preferred
Change view
Click to view available options
Menge:
30 μL
100 μL
Packungsgröße:
30 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17509842 100 μL -
17529842 30 μL 30 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17509842 Lieferant Origene Technologies Lieferanten-Nr. TA801475
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunogobulin genes. Forms a Schiff base with 5′-deoxyribose phosphate at abasic sites, but may not have lyase activity.
Spezifikation
Antigen POLI
Anwendungen Western Blot
Klassifikation Monoclonal
Klon OTI1H9
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gen POLI
Gen-Zugriffsnummer Q9UNA4
Gen-Alias RAD30B, RAD3OB, eta2
Gensymbole POLI
Wirtsspezies Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 458-740 of human POLI produced in E.coli.
Reinigungsverfahren Affinity Chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11201
Zielspezies Human
Inhalt und Lagerung -20° C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG1