missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2L Recombinant Rabbit Monoclonal Antibody (3C6H5)

Beschreibung
Immunogen sequence: MIIPVRCFTC GKIVGNKWEA YLGLLQAEYT EGDALDALGL KRYCCRRMLL AHVDLIEKLL NYAPLEK.
This gene encodes a subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains four conserved cysteines characteristic of an atypical zinc-binding domain. Like its counterpart in yeast, this subunit may be shared by the other two DNA-directed RNA polymerases.
Spezifikation
Spezifikation
| Antigen | POLR2L |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Recombinant Monoclonal |
| Klon | 3C6H5 |
| Konzentration | 1.14 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gen | POLR2L |
| Gen-Zugriffsnummer | P62875, P62876 |
| Gen-Alias | 2510029B14Rik; DNA-directed RNA polymerase III subunit L; DNA-directed RNA polymerases I, II, and III subunit RPABC5; hRPB7.6; hsRPB10b; Polr2l; polymerase (RNA) II (DNA directed) polypeptide L; polymerase (RNA) II (DNA directed) polypeptide L, 7.6kDa; polymerase (RNA) II subunit L; RBP10; RNA polymerase II 7.6 kDa subunit; RNA polymerase II subunit L; RNA polymerases I, II, and III subunit ABC5; RPABC5; RPB10; RPB10 homolog; RPB10beta; RPB7.6 |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?