Learn More
Chemsplash Polyethylene Disp Apron (Flat Pack) 70x115 cm
Beschreibung
10 Micron Disposable Apron
- Embossed Surface
- Ties At Waist
- Size:70x115cm
- Bag of 100
- PFAS Free
The 10 Micron Disposable Apron is a polyethylene single-use apron, ideally used in industrial kitchens, for washing or cleaning. This product is suitable for use in BRC-approved work areas and for the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Spezifikation
Spezifikation
| Typ | Apron |
| Größe | Universal |
| Material | Polyethylene |
| Farbe | Blue |
| Einwegprodukt | Single-use |
| Verschlusstyp | Waist Ties |
| Sterilität | Non-sterile |
| Latexfrei | Yes |
| Anzahl Taschen | None |
| Abmessungen (L x B) | 70 x 115 cm |
| Mehr anzeigen |