Learn More
Chemsplash Polypropylene Lab/Visitors Coat Navy with Velcro Closure
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
50 Stück
Beschreibung
Visitors Polypropylene Disposable Lab Coat
- Material: 30gsm Polypropylene Fabric
- Features:
- Velcro Closure
- With Fold Over Collar
- Elasticated Cuffs
- Ideal for Non-Hazardous Environments
- Ideal for Food Industry
- PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Spezifikation
Spezifikation
| Produkttyp | Visitor Lab Coat |
| Größe | Medium |
| Geschlecht | Unisex |
| Material | Polypropylene |
| Farbe | Navy |
| Verschlusstyp | Hook and Loop |
| Stulpenform | Elastic |
| Einwegprodukt | Single-use |
| Sterilität | Non-sterile |
| Widerstand | Particulates |
| Mehr anzeigen |