Learn More
Chemsplash Polypropylene Lab/Visitors Coat White with Velcro Closure
GREENER_CHOICE
Artikelnummer. 30058307
Click to view available options
Größe:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Packungsgröße:
50 Stück
Artikelnummer. 30058307
Marke: Chemsplash 2515.400.005
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Visitors Polypropylene Disposable Lab Coat with a velcro front closure. Suitable for visitors, site inspections or general day to day wear in minimal risk environments.
Visitors Polypropylene Disposable Lab Coat
  • Material: 30gsm Polypropylene Fabric
  • Features:
    • Velcro Closure
    • With Fold Over Collar
    • Elasticated Cuffs
    • Ideal for Non-Hazardous Environments
    • Ideal for Food Industry
    • PFAS Free
The Visitors Polypropylene Disposable Lab Coat is designed with a velcro front closure and is suitable for visitors, site inspections, or general wear in minimal risk environments. It is ideal for use in the food industry or other non-hazardous settings. This product is suitable for use in BRC-approved work areas and the pharmaceutical industry.
This product does not contain: PFAS, RoHS, POP, TSCA, REACH, or any conflict materials.
Spezifikation
Produkttyp Visitor Lab Coat
Größe Medium
Geschlecht Unisex
Material Polypropylene
Farbe White
Verschlusstyp Hook and Loop
Stulpenform Elastic
Einwegprodukt Single-use
Sterilität Non-sterile
Widerstand Particulates
Modell Knee-length
Anzahl Taschen None
Latexfrei Yes
Autoklavierbar Autoclavable