Learn More
Tocris Bioscience™ Pomalidomide 4'-alkylC5-azide
Beschreibung
Pomalidomide 4'-alkylC5-azide is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 4-[(5-Azidopentyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2434629-02-2 |
| Menge | 25 mg |
| Ziel | Degrader Building Blocks |
| Summenformel | C18H20N6O4 |
| Reinheit | 0.95 |