Learn More
Kaliumbromid, (kristallin/ACS-Reagenzgütegrad), MP Biomedicals
Artikelnummer. 11490542 Alle ansehen MP Biomedicals Produkte
Ansicht ändern
Click to view available options
Menge:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
11490542 100 g
11400552 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11490542 Lieferant MP Biomedicals Lieferanten-Nr. 0219142680
Please to purchase this item. Need a web account? Register with us today!
Chemische Identifikatoren
CAS 2-3-7758
Summenformel BrK
Molekulargewicht (g/mol) 119.00
MDL-Nummer MFCD00011358
InChI-Schlüssel IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC-Name Kaliumbromid
SMILES [K+].[Br-]
Spezifikation
Chemischer Name oder Material Potassium Bromide
Physikalische Form Crystalline
Menge 100 g
Formelmasse 119
Güte ACS-Reagenz