Learn More
Chemsplash PP Bouffant Mop Cap
Click to view available options
Farbe:
Blue
White
Packungsgröße:
1000 Stück
Beschreibung
Hand Made Polypropylene Mob Cap
- Material: 12GSM Polypropylene
- Hand Made
- Size: 21 in.
- With Elastic Band
- Pack of 100
- PFAS Free
Description: This product is suitable for use in BRC approved work areas and for the Pharmaceutical Industry. The hand-made mob cap is made from 12GSM polypropylene material and features an elastic band for a secure fit. It is PFAS-free and comes in a pack of 100, making it ideal for both food and pharmaceutical applications.
Spezifikation
Spezifikation
| Produkttyp | Bouffant |
| Größe | One Size |
| Material | Polypropylene |
| Farbe | White |
| Einwegprodukt | Single-use |
| Latexfrei | Yes |
| Sterilität | Non-sterile |