Learn More
Invitrogen™ Prasinezumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA558431
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Prasinezumab (PRX002/RG7935) is an investigational monoclonal antibody that targets alpha-synuclein, a protein that is believed to misfold and aggregate to form the protein structures that are highly implicated in Parkinson's disease pathology. Prasinezumab targets alpha-synuclein and is being developed through a worldwide collaboration with Roche as a potentially disease modifying therapy that is designed to slow or reduce the neurodegeneration associated with alpha-synuclein misfolding and/or its transmission.
Spezifikation
| Prasinezumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRX002; RG-7935 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |