missing translation for 'onlineSavingsMsg'
Learn More

pre-mRNA cleavage factor I (59 kDa subunit) Antibody, Alexa Fluor™ 750, Novus Biologicals™

Produktkod. 18552459 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Menge:
0.1 mL
Förpackningsstorlek:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Menge unitSize
18552459 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18552459 Leverantör Novus Biologicals Leverantörsnummer NB10061600AF750

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

pre-mRNA cleavage factor I (59 kDa subunit) Polyclonal antibody specifically detects pre-mRNA cleavage factor I (59 kDa subunit) in Human, Mammalia, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen pre-mRNA cleavage factor I (59 kDa subunit)
Anwendungen Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Alexa Fluor 750
Verdünnung Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Zusammensetzung 50mM Sodium Borate
Gen-Alias CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, CPSF 59 kDa subunit, FLJ12529,59 kDa subunit, FLJ39024, MGC9315, pre-mRNA cleavage factor I, 59 kDa subunit
Wirtsspezies Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human cleavage and polyadenylation specificity factor 7, 59 kDa subunit using the numbering given in entry NP_079087.2 (GeneID 79869).
Reinigungsverfahren Immunogen affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 79869
Zielspezies Human, Mammalia, Primate
Inhalt und Lagerung Store at 4C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.