missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pre-mRNA cleavage factor I (59 kDa subunit) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marke: Novus Biologicals NB100-61600
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
pre-mRNA cleavage factor I (59 kDa subunit) Polyclonal specifically detects pre-mRNA cleavage factor I (59 kDa subunit) in Human, Mammal, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Spezifikation
| pre-mRNA cleavage factor I (59 kDa subunit) | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CFIm59, cleavage and polyadenylation specific factor 7, 59kDa, CPSF 59 kDa subunit, FLJ12529,59 kDa subunit, FLJ39024, MGC9315, pre-mRNA cleavage factor I, 59 kDa subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79869 | |
| Human, Mammalia | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| NP_079087.2 | |
| CPSF7 | |
| The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human cleavage and polyadenylation specificity factor 7, 59 kDa subunit using the numbering given in entry NP_079087.2 (GeneID 79869). | |
| 100 μL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Dog, Horse, Guinea pig_10141, Panda, Orangutan, Rhesus Monkey, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat, Small-eared galago and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur