Learn More
Pregnenolonacetat, MP Biomedicals™
Artikelnummer. 11408390 Alle ansehen MP Biomedicals Produkte
Ansicht ändern
Click to view available options
Menge:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
11408390 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 11408390 Lieferant MP Biomedicals Lieferanten-Nr. 0210272001
Please to purchase this item. Need a web account? Register with us today!
Spezifikation
Chemischer Name oder Material Pregnenolone Acetate
CAS 1778-02-5
Summenformel C23H34O3
Menge 1 g
Synonym 1s,5s,10s,11s,14s,15s,2r-14-acetyl-2,15-dimethyltetracyclo 8.7.0.0<2,7>.0<11 ,15> heptadec-7-en-5-yl acetate, 11as-1-acetyl-9a,11a-dimethyl-1h,2h,3h,3ah,3bh,4h,6h,7h,8h,9h,9bh,10h,11h-cyclopenta a phenanthren-7-yl acetate, 13s-17-ethanoyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ethanoate, acetic acid 13s-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-yl ester
InChI-Schlüssel CRRKVZVYZQXICQ-BODHGXFRSA-N
SMILES CC(=O)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)OC(=O)C)C)C
IUPAC-Name [(13S)-17-Acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]acetat
Molekulargewicht (g/mol) 358.522
PubChem CID 18530247
Formelmasse 358.5
Physikalische Form Fest