Learn More
Liebherr™ Hochwertiger Gefrierschrank in Tischhöhe mit statischer Kühlung
Save space and energy. Liebherr™ Table-height Freezer with Static Cooling is an ultra-efficient unit that easily fits on or under a counter. Designed for better freezing performance and stable temperatures. Available with solid or wire basket drawers.
Marke:  Liebherr™ GGU 1500
Artikelnummer. 15355711
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen »
Beschreibung
Beschreibung
Optimal temperature stability and control
  • Large surface evaporator for better cooling performance and faster freezing of the interior contents
  • Highly efficient fan for consistent internal temperatures
  • Adjustable temperature control, down to -26°C
  • Digital temperature display with touch control buttons
  • Audible alarms for high temperatures and open door
Durable, user-friendly construction
  • Coated or stainless steel body with adjustable feet
  • Thermoformed polystyrene liner made from a single sheet for easy cleaning
  • Three 15cm grid or solid shelves, plus a 17cm bottom basket
  • Reversible, self-closing doors have interchangeable hinges for right or left open
  • Solid steel door lock for securing valuable samples and lab products
  • Ergonomic handle bar ensures effortless opening even when the door is frequently used
  • Highly energy efficient and uses eco-friendly refrigerant
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen