Learn More
Pritoxaximab Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Pritoxaximab Recombinant Monoclonal Antibody
Marke:  Invitrogen MA560165
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30284502
  • 477.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Pritoxaximab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Pritoxaximab
Recombinant Monoclonal
Unconjugated
caStx1; Shigamabs
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized