missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic-2,2-d2 Acid, CDN
Click to view available options
Menge:
5 g
Unit Size:
5 Gramm
Specifications
Specifications
| Chemischer Name oder Material | Propionic Acid 2,2-D2 |
| CAS | 19136-91-5 |
| Summenformel | C3 D2 H4 O2 |
| Menge | 5 g |
| Synonym | Propanoic-2,2-d2 acid (9 CI), Propionic-2,2-d2 acid (7 CI, 8 CI), α-Dideuteropropanolic acid, Propionic acid D2 (methylene-D2), Propionic acid 2,2-D2 |
| InChI-Formel | InChI=1 S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i2D2 |
| SMILES | [2 H]C([2 H])(C)C(=O)O |
| IUPAC-Name | 2,2-dideuteriopropanoic acid |
| Molekulargewicht (g/mol) | 76.091 |
| Formelmasse | 76.049 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction