missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prosurfactant Protein C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 539.00 €
Spezifikation
| Antigen | Prosurfactant Protein C |
|---|---|
| Verdünnung | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18435381
|
Novus Biologicals
NBP1-87201-25ul |
25 μL |
369.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18779673
|
Novus Biologicals
NBP1-87201 |
539.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
Prosurfactant Protein C Polyclonal specifically detects Prosurfactant Protein C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| Prosurfactant Protein C | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6440 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PEAQQRLALSEHLVTTATFSIGSTGLVVYDYQQLLIAYKPAPGTCCYIMKIAPESIPSLEALTRKVHNFQAKPAVPTSKLGQAEGRDAGSAPSGGDPAFLGMAVSTLCG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| PSP-C, pulmonary surfactant-associated protein C, Pulmonary surfactant-associated proteolipid SPL(Val), SFTP2, SFTP2pulmonary surfactant apoprotein-2 SP-C, SMDP2surfactant, pulmonary-associated protein C, SP-CSP5, surfactant protein C | |
| SFTPC | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts