Learn More
PRRX1 Antibody (CL14469) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-26855MFV610
Artikelnummer. 30293092
  • 836.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
PRRX1 Monoclonal antibody specifically detects PRRX1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
PRRX1
Monoclonal
mFluor Violet 610 SE
Homeobox protein PHOX1, paired mesoderm homeo box 1, paired mesoderm homeobox 1 isoform pmx-1b, paired mesoderm homeobox protein 1, paired related homeobox 1, Paired-related homeobox protein 1, PHOX1PRX-1, PMX1PRX1
This antibody was developed using a synthetic peptide derived from P54821, with the exact immunogen sequence remaining proprietary.
0.1 mL
Growth and Development, Neuronal Cell Markers
5396
Store at 4⪚C in the dark.
IgG2a
Western Blot
CL14469
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen