Learn More
PSD-95 Antibody (6G6-1C9) - BSA Free, Novus Biologicals™
Artikelnummer. 18751714 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
Artikelnummer. Menge unitSize
18751714 100 μL 100 Mikroliter
1 options
Artikelnummer. 18751714
Marke: Novus Biologicals NB300556
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
PSD-95 Monoclonal specifically detects PSD-95 in Human, Mouse, Rat, Invertebrate, Primate samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifica
Antigen PSD-95
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Klassifikation Monoclonal
Klon 6G6-1C9
Konzentration 3.3 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1:2000, Flow Cytometry 2 ug/ test (100ul), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Block/Neutralize
Gen-Alias discs, large homolog 4 (Drosophila), disks large homolog 4, FLJ97752, FLJ98574, post-synaptic density protein 95, Postsynaptic density protein 95, PSD-95, PSD95SAP90SAP-90discs large homolog 4, Synapse-associated protein 90, Tax interaction protein 15
Gensymbole DLG4
Wirtsspezies Mouse
Immunogen Purified recombinant rat PSD-95.
Reinigungsverfahren Protein A purified
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Neuronal Cell Markers, Neuroscience, Neurotransmission
Primär oder sekundär Primary
Gen-ID (Entrez) 1742
Testspezifität Detects Post Synaptic Density 95 kDa (PSD-95) from rat tissues.
Zielspezies Human, Mouse, Rat, Invertebrate, Primate
Inhalt und Lagerung Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Vedi altri risultati Mostra meno risultati
For Research Use Only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.