missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSEN2 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5148553
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1 or PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor such that, they either directly regulate gamma-secretase activity, or themselves act are protease enzymes. Two alternatively spliced transcript variants encoding different isoforms of PSEN2 have been identified.
Spezifikation
| PSEN2 | |
| Polyclonal | |
| Unconjugated | |
| PSEN2 | |
| AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5664 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P49810 | |
| PSEN2 | |
| E. coli-derived recombinant human Presenilin-2 C-Terminal Fragment, Glu322-Thr388. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur