missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSG6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA598094
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
PSG6 Polyclonal Antibody for Western Blot, ELISA
The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
Spezifikation
| PSG6 | |
| Polyclonal | |
| Unconjugated | |
| PSG6 | |
| CGM3; pregnancy specific beta-1-glycoprotein 6; pregnancy-specific beta-1-glycoprotein 10; Pregnancy-specific beta-1-glycoprotein 12; pregnancy-specific beta-1-glycoprotein 6; PS-beta-G-10; PS-beta-G-12; PS-beta-G-6; PSBG-10; PSBG-12; PSBG-6; PSG10; PSG12; PSG6; PSGGB | |
| Rabbit | |
| Protein G | |
| RUO | |
| 5675 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q00889 | |
| PSG6 | |
| Recombinant Human Pregnancy-specific beta-1-glycoprotein 6 protein (35-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur