missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PSMA1 Recombinant Rabbit Monoclonal Antibody (24GB1610)

Beschreibung
PSMA1 Recombinant Monoclonal Antibody for Western Blot, Flow
Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signaling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (approximately31), barrel-shaped molecular machine with an apparent molecular weight of approximately2. 5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.
Spezifikation
Spezifikation
| Antigen | PSMA1 |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB1610 |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | PSMA1 |
| Gen-Zugriffsnummer | P18420, P25786, Q9R1P4 |
| Gen-Alias | 20S proteasome subunit C2; 30 kDa prosomal protein; alpha-type; C2; epididymis secretory protein Li 275; HC2; HEL-S-275; macropain subunit C2; macropain subunit nu; MGC14542; MGC14575; MGC14751; MGC1667; MGC21459; MGC22853; MGC23915; multicatalytic endopeptidase complex subunit C2; NU; PROS30; PROS-30; proteasome (prosome, macropain) subunit, alpha type 1; proteasome (prosome, macropain) subunit, alpha type, 1; proteasome 20S subunit alpha 1; proteasome alpha 1 subunit; proteasome component C2; Proteasome nu chain; proteasome subunit alpha 1; Proteasome subunit alpha type-1; proteasome subunit nu; proteasome subunit, alpha-type, 1; protein P30-33K; PSC2; Psma1; testicular tissue protein Li 150 |
| Gensymbole | PSMA1 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?