Learn More
Bohlender™ Bola-Schliff-GL-3-Wege-Hähne GL18 NS 29/32 Drm. 6m
Artikelnummer. 15307792
Ansicht ändern
Click to view available options
Material:
PTFE
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Material Packungsgröße
15307792 PTFE 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15307792 Lieferant Bohlender™ Lieferanten-Nr. E69018
  • ONLINE EXCLUSIVE PRICE
    301.51 €
    172.72 € / 1 Stück
    Sparen 128.79 €
    42% Rabatt
Verfügbar ab: 16-11-2026