missing translation for 'onlineSavingsMsg'
Learn More

PTH Antibody (SPM604), Alexa Fluor™ 488 - N-terminal, Novus Biologicals™

Artikelnummer. 18441624 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μg
Unit Size:
100 Mikroliter
Product Code. Menge unitSize
18441624 100 μg 100 Mikroliter
1 options
This item is not returnable. View return policy

Product Code. 18441624

Brand: Novus Biologicals NBP254434AF488

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PTH Monoclonal antibody specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon SPM604
Konjugat Alexa Fluor 488
Zusammensetzung 50 mM Sodium Borate
Gen-Alias Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1
Wirtsspezies Mouse
Immunogen A synthetic peptide around aa 1-34 of human mature-PTH-polypeptide (exact sequence is proprietary) (Uniprot: P01270)
Reinigungsverfahren Protein A or G purified
Menge 100 μg
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, BiologicAll speciesy Active Proteins, Cancer
Primär oder sekundär Primary
Gen-ID (Entrez) 5741
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.