Learn More
Saint-Gobain Performance Plastics PureFlo™ Z Series PES Mini Capsules
Artikelnummer. p-200132120
Click to view available options
Porengröße:
0.2 μm
0.45 μm
0.45/0.2 μm
Packungsgröße:
30 Stück
Artikelnummer. 30428857
Marke: Saint-Gobain Performance Plastics MZPZS020D2H2H
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Hydrophilic, gamma-irradiatable filtration units designed for single-use sterile assemblies. With superior flow, throughput, broad chemical compatibility and scalable. They are reliable pre- and post-use integrity testing for critical biopharmaceutical processes.
  • PureFlo™ Z Series PES Capsules are designed for single-use sterile assemblies, providing hydrophilic filtration for critical biopharmaceutical processes.
  • Gamma-irradiatable design supports effective sterilization, ensuring compatibility with stringent pharmaceutical requirements.
  • Superior throughput capacity and flow performance enhance operational efficiency in liquid applications.
  • Hydrophilic, asymmetric polyethersulfone (PES) membrane delivers reliable filtration and chemical compatibility across a broad range of substances.
  • Available with a final sterilizing grade 0.1 or 0.2 μm membrane, ensuring effective filtration.
  • Optional built-in ZenFlo™ prefilter enhances performance and efficiency.
  • Carefully selected materials and design enable quick and reliable pre- and post-use integrity testing for critical applications.
  • Scalable size options simplify scale-up throughout all stages of drug development and manufacturing processes.
  • Autoclavable construction ensures flexibility in sterilization methods for optimized usability.
  • Lightweight and compact design integrates seamlessly into biopharmaceutical assemblies without adding bulk.
  • Promotes optimal process efficiency, minimizing downtime and supporting productivity in demanding environments.
TRUSTED_SUSTAINABILITY
Spezifikation
Einwegprodukt Single-use
Filtertyp Filter Capsule
Material (Membran) Hydrophilic PES
Porengröße 0.45/0.2 μm
Sterilität Non-sterile
Typ Mini Capsules