Learn More
PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle
Used in preparing small samples for pathological and biochemical studies
Marke: PYREX™ 9651694
Benachrichtigungen:
If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus from pieces of connective tissue or muscle. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.
Beschreibung
Homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward. Mortar is unground and pestle contact surface is PTFE threaded onto a stainless steel shaft.
Caution: If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.
Bestellinformation
Sold in a Case of 1.
Spezifikation
| Non-sterile | |
| Borosilicate Glass | |
| Preparing small samples for pathological and biochemical studies | |
| 7.08 in. | |
| 0.9 in. | |
| 25 mm | |
| PTFE |
| 17 mL | |
| Potter-Elvehjem tissue grinder | |
| Case of 1 | |
| 180 mm | |
| Tooled mouth | |
| 0.57 oz., 17 mL |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.