missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridin-4-carbonitril, Mikromol™
Beschreibung
Pyridin-4-carbonitril, Mikromol
Spezifikation
Specifications
| Chemischer Name oder Material | Pyridine-4-carbonitrile |
| CAS | 100-48-1 |
| Bezeichnung der Analyten oder Bestandteile | Pyridine-4-carbonitrile |
| Versandbedingung | Raumtemperatur |
| Summenformel | C6 H4 N2 |
| Zertifizierung/Konformität | ISO 9001 |
| Synonym | 4-Pyridinecarbonitrile, Isonicotinonitrile (6CI,8CI), 4-Azabenzonitrile, 4-Cyanopyridine, 4-Pyridinenitrile, 4-Pyridyl cyanide, 4-Pyridylcarbonitrile, Isonicotinic acid nitrile, NSC 60681, gamma-Cyanopyridine, Fampridine Impurity 2, Ethionamide Impurity B, Isoniazid Impurity C |
| InChI-Formel | InChI=1S/C6H4N2/c7-5-6-1-3-8-4-2-6/h1-4H |
| Lösungstyp | Unverdünnt |
| SMILES | N#Cc1ccncc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?